missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Density Determination Kit

Product Code. 17957320 Shop All Sartorius Products
Change view
Click to view available options
:
Type: Density Determination Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17957320 Type: Density Determination Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17957320 Supplier Sartorius Supplier No. YDK03MC

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Density Determination Kit for Cubis™ II

  • Cubis™ II Accessory
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Cubis™ II High-Capacity Micro Balance
Includes Density Determination Set for Solids and Liquids
Type Density Determination Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.