missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DENR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554507
This item is not returnable.
View return policy
Description
Immunogen sequence: AADISESSGA DCKGDPRNSA KLDADYPLRV LYCGVCSLPT EYCEYMPDVA KCRQWLEKNF PNEFAKLTVE NSPKQEAGIS EGQGTAGEEE EKK Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 86%.
This gene encodes a protein whose expression was found to increase in cultured cells at high density but not during growth arrest. This gene was also shown to have increased expression in cells overexpressing HER-2/neu proto-oncogene. The protein contains an SUI1 domain. In budding yeast, SUI1 is a translation initiation factor that along with eIF-2 and the initiator tRNA-Met, directs the ribosome to the proper translation start site. Proteins similar to SUI have been found in mammals, insects, and plants.
Specifications
| DENR | |
| Polyclonal | |
| Unconjugated | |
| DENR | |
| 1500003K04Rik; Denr; density regulated re-initiation and release factor; density-regulated protein; DRP; DRP1; H14; Protein DRP1; SMAP-3; smooth muscle cell associated protein-3; smooth muscle cell-associated protein 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8562 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43583 | |
| DENR | |
| Recombinant protein corresponding to Human DENR. Recombinant protein control fragment (Product #RP-92545). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction