missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DENND4C Mouse anti-Human, Mouse, Rat, Clone: 300000000, Novus Biologicals™
Shop All Bio Techne ProductsDescription
DENND4C Monoclonal specifically detects DENND4C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
Specifications
| Antigen | DENND4C |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 300000000 |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Formulation | PBS (pH 7.4), 50% Glycerol |
| Gene Alias | bA513M16.3, C9orf55B, DENN/MADD domain containing 4C, DKFZp686I09113, FLJ20686 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?