missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DENND4C Mouse anti-Human, Mouse, Rat, Clone: 300000000, Novus Biologicals™
Mouse Monoclonal Antibody
£195.00 - £447.00
Specifications
| Antigen | DENND4C |
|---|---|
| Clone | 300000000 |
| Concentration | 1 mg/ml |
| Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18325413
|
Novus Biologicals
NBP3-11040-25UG |
25 μg |
£195.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18354705
|
Novus Biologicals
NBP3-11040-100UG |
100 μg |
£447.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DENND4C Monoclonal specifically detects DENND4C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| DENND4C | |
| 1 mg/ml | |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| bA513M16.3, C9orf55B, DENN/MADD domain containing 4C, DKFZp686I09113, FLJ20686 | |
| Full Length Human Recombinant DENND4C protein (NM_017925) | |
| Primary | |
| Store at -20°C. |
| 300000000 | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 50% Glycerol | |
| 55667 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title