missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DENND4C Monoclonal Antibody (3E8), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545961
This item is not returnable.
View return policy
Description
A 1:1,000 dilution of MA5-45961 was sufficient for detection of DENND4C in A549, MEF cells by Western Blot analysis. Detects approximately 213 kDa.
DENND4C is guanine nucleotide exchange factor (GEF) activating RAB10. It promotes the exchange of GDP to GTP, converting inactive GDP-bound RAB10 into its active GTP-bound form. Thereby, stimulates SLC2A4/GLUT4 glucose transporter-enriched vesicles delivery to the plasma membrane in response to insulin.
Specifications
| DENND4C | |
| Monoclonal | |
| 1 mg/mL | |
| PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4 | |
| A6H8H2, Q5VZ89 | |
| DENND4C | |
| Full Length Human Recombinant Dennd4c protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 3E8 | |
| FITC | |
| DENND4C | |
| 1700065A05Rik; AA420392; bA513M16.3; C9orf55; C9orf55B; DENN domain containing 4C; DENN domain-containing protein 4C; DENN domain-containing protein 4C-like protein; DENN/MADD domain containing 4A pseudogene; DENN/MADD domain containing 4C; Dennd4c; likely ortholog of H. sapiens chromosome 9 open reading frame 55 (C9orf55); RAB10GEF; RGD1308489 | |
| Mouse | |
| Protein G | |
| RUO | |
| 313340, 329877, 55667 | |
| 4°C, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction