missing translation for 'onlineSavingsMsg'
Learn More

DENND4C Antibody (3E8), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18501745 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18501745 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18501745 Supplier Novus Biologicals Supplier No. NBP311040AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DENND4C Monoclonal antibody specifically detects DENND4C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DENND4C
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 300000000
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias bA513M16.3, C9orf55B, DENN/MADD domain containing 4C, DKFZp686I09113, FLJ20686
Host Species Mouse
Immunogen Full Length Human Recombinant DENND4C protein (NM_017925)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cellular Signaling, Cytokines and Growth Factors, Membrane Trafficking and Chaperones, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 55667
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.