Learn More
Invitrogen™ DENND4A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120423
Description
Positive test controls include: HepG2, mouse liver. The target is usually found in the following locations: Nucleus. Immunogen sequence: RPGRYFLKSS PSTENMHFPS SISSQTRQSC ISTSASGLDT SALSVQGNFD LNSKSKLQEN FCTRSIQIPA NRSKTAMSKC PIFPMARSIS TSGPLDKEDT GRQKLISTGS LPATLQGATD SLGLEWHLPS PDPVTVPYLS PLVVWKELES L.
Probable guanine nucleotide exchange factor (GEF) which may activate RAB10. Promotes the exchange of GDP to GTP, converting inactive GDP-bound Rab proteins into their active GTP-bound form. According to PubMed:8056341, it may bind to ISRE-like element (interferon-stimulated response element) of MYC P2 promoter.
Specifications
| DENND4A | |
| Polyclonal | |
| Unconjugated | |
| DENND4A | |
| AI115600; AI851943; c-myc promoter binding protein; C-myc promoter-binding protein; DENN domain containing 4A; DENN domain-containing protein 4A; DENN/MADD domain containing 4A; DENND4A; F730015K02Rik; IRLB; MYCPBP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 102442, 10260 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.45 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q7Z401 | |
| DENND4A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1500-1650 of human DENND4A (NP_005839.3). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.