missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DENND2C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112989
This item is not returnable.
View return policy
Description
DENND2C Polyclonal Antibody for ELISA, Western Blot
Guanine nucleotide exchange factor (GEF) which may activate RAB9A and RAB9B. Promotes the exchange of GDP to GTP, converting inactive GDP-bound Rab proteins into their active GTP-bound form.
Specifications
| DENND2C | |
| Polyclonal | |
| Unconjugated | |
| DENND2C | |
| A930010I20Rik; DENN domain containing 2C; DENN domain-containing protein 2C; DENN/MADD domain containing 2C; DENND2C; dJ1156J9.1; RGD1308197; RP5-1156J9.1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 163259 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q68D51 | |
| DENND2C | |
| Recombinant Human DENN domain-containing protein 2C protein (1-300AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction