missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DENND1A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25658-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody (25658-1-AP) can recognize all isoforms at 112 kDa and 52-64 kDa.
Clathrin-mediated endocytosis is a major mechanism for internalization of proteins and lipids. Members of the connecdenn family, such as DENND1A, function as guanine nucleotide exchange factors (GEFs) for the early endosomal small GTPase RAB35 (MIM 604199) and bind to clathrin and clathrin adaptor protein-2 (AP2). Thus, connecdenns link RAB35 activation with the clathrin machinery.Specifications
| DENND1A | |
| Polyclonal | |
| Unconjugated | |
| DENND1A | |
| Connecdenn, DENN/MADD domain containing 1A, DENND1A, FAM31A, KIAA1608, Protein FAM31A, RP11 230L22.3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57706 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8TEH3 | |
| DENND1A | |
| DENND1A Fusion Protein Ag22547 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction