missing translation for 'onlineSavingsMsg'
Learn More

DENND1A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18382196 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382196 100 μL 100µL
18317476 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18382196 Supplier Novus Biologicals Supplier No. NBP304911100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DENND1A Polyclonal specifically detects DENND1A in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DENND1A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias Connecdenn, Connecdenn 1, DENN/MADD domain containing 1A, FAM31ADENN domain-containing protein 1A, FLJ21129, FLJ38464, KIAA1608connecdenn, Protein FAM31A, RP11-230L22.3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 270-527 of human DENND1A (NP_079096.2).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57706
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.