missing translation for 'onlineSavingsMsg'
Learn More

Dengue Virus NS5 Antibody (GT353), Novus Biologicals™

Product Code. 18332375 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18332375 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18332375 Supplier Novus Biologicals Supplier No. NBP313489

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dengue Virus NS5 Monoclonal antibody specifically detects Dengue Virus NS5 in Virus samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dengue Virus NS5
Applications Western Blot, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone GT353
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunofluorescence 1:100-1:2000, Immunoprecipitation
Formulation PBS
Gene Alias Dengue NS5, Dengue NS5 protein
Host Species Mouse
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Dengue Virus NS5 (Dengue virus 2 (strain 16681 PDK 53)). The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1489713
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.