missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus NS1 protein Antibody, Alexa Fluor™ 750, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Dengue Virus NS1 protein Polyclonal antibody specifically detects Dengue Virus NS1 protein in Virus samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | Dengue Virus NS1 protein |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 750 |
| Formulation | 50mM Sodium Borate |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Dengue virus (DENV) (type 2, strain New Guinea C) NS1 (AAC59275.1; Asp1-Ala352). |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?