missing translation for 'onlineSavingsMsg'
Learn More

Dengue Virus Capsid Protein Antibody (GT574), Novus Biologicals™

Product Code. 18341412 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341412 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341412 Supplier Novus Biologicals Supplier No. NBP313551

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dengue Virus Capsid Protein Monoclonal antibody specifically detects Dengue Virus Capsid Protein in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dengue Virus Capsid Protein
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GT574
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS
Gene Alias polyprotein gene
Host Species Mouse
Immunogen Full length Dengue virus Capsid recombinant protein. (Dengue virus 2 (strain 16681 PDK 53))
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1494449
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.