Learn More
Dengue Virus Antibody (Dengue 1-11(3)), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-65685R
Product Code. 18072106
  • £484.00 / 125µL
Description
Dengue Virus Monoclonal specifically detects Dengue Virus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dengue Virus
Monoclonal
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
Protein A purified
RUO
Primary
Virus
Purified
Western Blot, ELISA, Immunohistochemistry (Paraffin)
Dengue 1-11(3)
Western Blot, ELISA, Immunohistochemistry-Paraffin
DEN 1, DEN1, Dengue hemorrhagic fever, Dengue type 1 virus, DENV 1, DENV1
Made to Viral DENGUE VIRUS
0.125 mL
Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
NB100-65685 recognizes all four dengue virus serotypes (DEN-1, DEN-2, DEN-3 and DEN-4 least strongly) of the genus Flavivirus. NB100-65685 detects an anti-E/envelope specific band of 61kDa under reducing conditions in Western blotting. A weak secondary band of 80kDa may also be apparent.
Store at 4C in the dark.
IgG2a