missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus Antibody (Dengue 1-11(3)), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65685AF750
This item is not returnable.
View return policy
Description
Dengue Virus Monoclonal antibody specifically detects Dengue Virus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Dengue Virus | |
| Monoclonal | |
| Alexa Fluor 750 | |
| DEN 1, DEN1, Dengue hemorrhagic fever, Dengue type 1 virus, DENV 1, DENV1 | |
| Made to Viral DENGUE VIRUS | |
| 0.125 mL | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| Virus | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Dengue 1-11(3) | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction