missing translation for 'onlineSavingsMsg'
Learn More

Dengue Virus 4 NS1 Antibody (971314) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30763209 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30763209 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30763209 Supplier Novus Biologicals Supplier No. FAB9444JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dengue Virus 4 NS1 Monoclonal antibody specifically detects Dengue Virus 4 NS1 in Virus samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dengue Virus 4 NS1
Applications Western Blot
Classification Monoclonal
Clone 971314
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias Dengue NS1, Dengue pan
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived recombinant Dengue Virus 4 NS1 protein, Asp1-Ala352, Accession # NP_740318
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.