missing translation for 'onlineSavingsMsg'
Learn More

Dengue Virus 3 NS1 Mouse anti-Virus, Clone: CC6.BC5.D8, Novus Biologicals™

Product Code. 18355502 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.1mg
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355502 0.5 mg 0.5mg
18309984 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18355502 Supplier Bio-Techne Supplier No. NBP3148450.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dengue Virus 3 NS1 Monoclonal antibody specifically detects Dengue Virus 3 NS1 in Virus samples. It is validated for ELISA, ELISA, Direct ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dengue Virus 3 NS1
Applications ELISA
Classification Monoclonal
Clone CC6.BC5.D8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Sandwich ELISA, Direct ELISA
Formulation PBS pH 7.4
Host Species Mouse
Immunogen Recombinant Dengue virus NS1, a mixture of serotypes 1-4
Purification Method Protein G purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.