missing translation for 'onlineSavingsMsg'
Learn More

Dengue Virus 2 VLP/Envelope Mouse anti-Virus, Clone: ME1.G6.C12, Novus Biologicals™

Product Code. 18339675 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.10mg
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339675 0.5 mg 0.5mg
18333674 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18339675 Supplier Novus Biologicals Supplier No. NBP3148430.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dengue Virus 2 VLP/Envelope Monoclonal antibody specifically detects Dengue Virus 2 VLP/Envelope in Virus samples. It is validated for ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dengue Virus 2 VLP/Envelope
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone ME1.G6.C12
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS pH 7.4
Gene Alias Envelope protein E
Host Species Mouse
Immunogen Recombinant Dengue virus NS1, a mixture of serotypes 1-4
Purification Method Protein G purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.