missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus 2 premembrane (prM) Envelope Mouse anti-Virus, Clone: CC5.A9.E10, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Dengue Virus 2 premembrane (prM) Envelope Monoclonal antibody specifically detects Dengue Virus 2 premembrane (prM) Envelope in Virus samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | Dengue Virus 2 premembrane (prM) Envelope |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | CC5.A9.E10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | PBS pH 7.4 |
| Host Species | Mouse |
| Immunogen | Recombinant Dengue virus Virus-like particles, a mixture of serotypes 1-4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?