missing translation for 'onlineSavingsMsg'
Learn More

Dengue Virus 2 NS1 Mouse anti-Virus, Clone: LH5.H7.B12.D7, Novus Biologicals™

Product Code. 18388594 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.10mg
0.50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18388594 0.1 mg 0.10mg
18399985 0.5 mg 0.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18388594 Supplier Novus Biologicals Supplier No. NBP3148420.1MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dengue Virus 2 NS1 Monoclonal antibody specifically detects Dengue Virus 2 NS1 in Virus samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dengue Virus 2 NS1
Applications ELISA
Classification Monoclonal
Clone LH5.H7.B12.D7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Sandwich ELISA
Formulation PBS pH 7.4
Gene Alias Dengue NS1, Dengue NS1 protein, Dengue Virus
Host Species Mouse
Immunogen Recombinant Dengue virus NS1, a mixture of serotypes 1-4
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.