missing translation for 'onlineSavingsMsg'
Learn More

Dengue Virus 2 Envelope Antibody, HRP, Novus Biologicals™

Product Code. 18351164 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18351164 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18351164 Supplier Novus Biologicals Supplier No. NBP305839H

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Dengue Virus 2 Envelope Polyclonal specifically detects Dengue Virus 2 Envelope in Virus samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dengue Virus 2 Envelope
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Dengue virus (DENV) (type 2, strain New Guinea C/PUO-218 hybrid) E/ Envelope Protein (AAC59274.1; Met578-Gln680).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.