Learn More
Invitrogen™ Dendritic Cell Marker DCIR2 Monoclonal Antibody (33D1), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen™ 14-5884-81
Description
Description: The 33D1 monoclonal antibody reacts with a mouse dendritic cell (DC)-specific surface marker, DCIR2. The nature and biological activity of DCIR2 is unknown. DCIR2 has been reported on a variety of dendritic cell subpopulations from mouse thymus, spleen, lymph node, and Peyer's patch. Bone marrow dendritic cells require GM-CSF to express DCIR2, and this expression is downregulated in the presence of IL-4. DCIR2 has been detected in vivo in brain dendritic cells post infection with Toxoplasma gondii. Applications Reported: This 33D1 antibody has been reported for use in flow cytometric analysis, immunohistology staining of frozen tissue sections, and immunohistochemical staining. It has also been reported in complement-mediated cytotoxicity. Applications Tested: The 33D1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. For detection of purified 33D1 it is recommended not to use goat anti-rat due to its low affinity for 33D1. Rather, we recommend using biotin mouse anti-rat (cat. 13-4813).
The nature and biological activity of DCIR2 is unknown. DCIR2 has been reported on a variety of dendritic cell subpopulations from mouse thymus, spleen, lymph node, and Peyer's patch. Bone marrow dendritic cells require GM-CSF to express DCIR2, and this expression is downregulated in the presence of IL-4. DCIR2 has been detected in vivo in brain dendritic cells post infection with Toxoplasma gondii.
Specifications
| Dendritic Cell Marker DCIR2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 0 | |
| Clec4a4 | |
| Affinity chromatography | |
| RUO | |
| 474145 | |
| 4°C | |
| Liquid |
| Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen) | |
| 33D1 | |
| Unconjugated | |
| Clec4a4 | |
| Clec4a4; C-type lectin domain family 4, member a4; DC Marker; Dcir2; dendritic cell inhibitory receptor 2 | |
| Rat | |
| 50 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.