missing translation for 'onlineSavingsMsg'
Learn More

Demeton-S Sulfoxide, TRC

Product Code. 30137887
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137887 25 mg 25mg
18060550 10 mg 10mg
18034040 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137887 Supplier Toronto Research Chemicals Supplier No. TRCD23139025MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2496-92-6
Molecular Formula C8 H19 O4 P S2
Molecular Weight (g/mol) 274.34
Synonym Phosphorothioic acid, O,O-diethyl S-[2-(ethylsulfinyl)ethyl] ester, Ethanethiol, 2-(ethylsulfinyl)-, S-ester with O,O-diethyl phosphorothioate (8CI), Demeton-S sulfoxide, Isosystox sulfoxide, O,O-Diethyl S-[2-(ethylsulfinyl)ethyl] phosphorothioate, Systox sulfoxide
IUPAC Name 1-diethoxyphosphorylsulfanyl-2-ethylsulfinylethane
SMILES CCOP(=O)(OCC)SCCS(=O)CC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C8H19O4PS2/c1-4-11-13(9,12-5-2)14-7-8-15(10)6-3/h4-8H2,1-3H3
Formula Weight 274.0462
Percent Purity >95%
Chemical Name or Material Demeton-S Sulfoxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.