missing translation for 'onlineSavingsMsg'
Learn More

Demeton-S-methyl, TRC

Product Code. 17909769
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17909769 50 mg 50mg
18071450 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17909769 Supplier Toronto Research Chemicals Supplier No. TRCD23137050MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 919-86-8
Molecular Formula C6 H15 O3 P S2
Molecular Weight (g/mol) 230.29
Synonym Phosphorothioic acid, S-[2-(ethylthio)ethyl] O,O-di-Me ester (6CI), Demeton-S-methyl, Metaisoseptox, Metaisosystox, Metasystox 55, Metasystox I, Metasystox J, Methyl demeton thioester, Methyl isosystox, Methylthionodemeton, O,O-Dimethyl S-2-(ethylthio)ethyl phosphorothioate, S-[2-(Ethylthio)ethyl] O,O-dimethyl phosphorothioate, S-[2-(Ethylthio)ethyl] O,O-dimethyl thiophosphate, Thiometon oxon
IUPAC Name 1-dimethoxyphosphorylsulfanyl-2-ethylsulfanylethane
SMILES CCSCCSP(=O)(OC)OC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H15O3PS2/c1-4-11-5-6-12-10(7,8-2)9-3/h4-6H2,1-3H3
Formula Weight 230.02
Percent Purity >95%
Chemical Name or Material Demeton-S-methyl
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.