Product Code. 30367263
Click to view available options
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
unitSize
30367263
50 mg
50mg
30367012
5 mg
5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
30367263
Supplier
Toronto Research Chemicals
Supplier No.
TRCD23136250MG
Please sign in to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Chemical Identifiers
Molecular Formula
C8H9D10O3PS2
Molecular Weight (g/mol)
268.4
Synonym
O,O-Diethyl S-[2-(Ethylthio)ethyl]phosphorothioate-d10, Thioldemeton-d10, Demetonthiol-d10, Systox-thiol-d10, Isosystox-d10
SMILES
[2H]C([2H])([2H])C([2H])([2H])OP(=O)(OC([2H])([2H])C([2H])([2H])[2H])SCCSCC
Specifications
Quantity
50 mg
InChI Formula
InChI=1S/C8H19O3PS2/c1-4-10-12(9,11-5-2)14-8-7-13-6-3/h4-8H2,1-3H3/i1D3,2D3,4D2,5D2
Chemical Name or Material
Demeton-S-d10
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?