missing translation for 'onlineSavingsMsg'
Learn More

Demeton-O-methyl, TRC

Product Code. 30137381
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137381 500 mg 500mg
30135839 250 mg 250mg
30007240 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137381 Supplier Toronto Research Chemicals Supplier No. TRCD231363500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 867-27-6
Molecular Formula C6 H15 O3 P S2
Molecular Weight (g/mol) 230.2853
Synonym Phosphorothioic acid, O-[2-(ethylthio)ethyl] O,O-dimethyl ester, Phosphorothioic acid, O-[2-(ethylthio)ethyl] O,O-di-Me ester (6CI), Demeton-O-methyl, Methyl-O-demeton, O,O-Dimethyl O-2-(ethylthio)ethyl phosphorothioate, O,O-Dimethyl O-[2-(ethylthio)ethyl] thiophosphate, O-[2-(Ethylthio)ethyl] O,O-dimethyl phosphorothioate, O-[2-(Ethylthio)ethyl] O,O-dimethyl thiophosphate
IUPAC Name 2-ethylsulfanylethoxy-dimethoxy-sulfanylidene-λ^{5}-phosphane
SMILES CCSCCOP(=S)(OC)OC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H15O3PS2/c1-4-12-6-5-9-10(11,7-2)8-3/h4-6H2,1-3H3
Formula Weight 230.02
Percent Purity >95%
Chemical Name or Material Demeton-O-methyl
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.