missing translation for 'onlineSavingsMsg'
Learn More

Demeton-O-methyl Sulfone, TRC

Product Code. 30363204
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30363204 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30363204 Supplier Toronto Research Chemicals Supplier No. TRCD231361500MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Demeton-O-methyl Sulfone
CAS 25476-48-6
Molecular Formula C6 H15 O5 P S2
Quantity 500 mg
Synonym O-[2-(Ethylsulfonyl)ethyl] O,O-Dimethyl Phosphorothioate, Disulfoton Oxygen Analog Sulfone
InChI Formula InChI=1S/C6H15O5PS2/c1-4-14(7,8)6-5-11-12(13,9-2)10-3/h4-6H2,1-3H3
SMILES CCS(=O)(=O)CCOP(=S)(OC)OC
IUPAC Name 2-ethylsulfonylethoxy-dimethoxy-sulfanylidene-λ^{5}-phosphane
Molecular Weight (g/mol) 262.284
Formula Weight 262.01
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.