missing translation for 'onlineSavingsMsg'
Learn More

Demeclocycline hydrochloride

Product Code. 15455369
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15455369 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15455369 Supplier Thermo Scientific Alfa Aesar Supplier No. J63102.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 64-73-3 | C21H22Cl2N2O8 | 501.313 g/mol

Demeclocycline hydrochloride is used as an antibiotic related to tetracycline and produced by streptomyces aureofaciens, and used to maintain effective blood levels for longer periods of time, used as a fluorescent bone-labeling agent in bone histomorphometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Demeclocycline hydrochloride is used as an antibiotic related to tetracycline and produced by streptomyces aureofaciens, and used to maintain effective blood levels for longer periods of time, used as a fluorescent bone-labeling agent in bone histomorphometry.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-73-3
Molecular Formula C21H22Cl2N2O8
Molecular Weight (g/mol) 501.313
MDL Number MFCD00082371
InChI Key OAPVUSSHCBRCOL-KBHRXELFSA-N
Synonym demeclocycline hydrochloride, demeclocycline hcl, declomycin, detravis, ledermycine, clortetrin, ledermycin, ledermicina, veraciclina, demeplus
PubChem CID 54686764
IUPAC Name (4S,4aS,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrochloride
SMILES CN(C)C1C2CC3C(C4=C(C=CC(=C4C(=C3C(=O)C2(C(=C(C1=O)C(=O)N)O)O)O)O)Cl)O.Cl

Specifications

Melting Point >245°C (decomposition)
Color Yellow
Quantity 1 g
Beilstein 3849198
Merck Index 14,2886
Solubility Information Soluble in water.
Formula Weight 501.32
Physical Form Powder
Chemical Name or Material Demeclocycline hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.