missing translation for 'onlineSavingsMsg'
Learn More

Demeclocycline Hydrochloride, Mikromol™

Product Code. 17972456
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17972456 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17972456 Supplier Mikromol Supplier No. MM0636.00
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Demeclocycline Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Demeclocycline Hydrochloride
CAS 64-73-3
Analyte or Component Names Demeclocycline Hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C21 H21 Cl N2 O8 . Cl H
Certifications/Compliance ISO 9001
Synonym Chlortetracycline Hydrochloride Imp. B (EP), Chlortetracycline Imp. B (EP), Demeclocycline Hydrochloride, Chlortetracycline Hydrochloride Imp. B (EP) as Hydrochloride, 7-Chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride, (4S,4aS,5aS,6S,12aS)-7-Chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride, [4S-(4alpha,4aalpha,5aalpha,6beta,12aalpha)]-7-Chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride, 6-Demethyl-7-chlorotetracycline hydrochloride, 7-Chloro-6-demethyltetracycline hydrochloride, Clortetrin, Declomycin, Declomycin hydrochloride, Demeclocycline hydrochloride, Demethylchlorotetracycline hydrochloride, Demetraciclina, Detravis, Ledermycin hydrochloride, Meciclin, Chlortetracycline Hydrochloride Impurity B as Hydrochloride, Chlortetracycline Impurity B as Hydrochloride
InChI Formula InChI=1S/C21H21ClN2O8.ClH/c1-24(2)14-7-5-6-10(16(27)12-9(25)4-3-8(22)11(12)15(6)26)18(29)21(7,32)19(30)13(17(14)28)20(23)31;/h3-4,6-7,14-15,25-26,28-29,32H,5H2,1-2H3,(H2,23,31);1H/t6-,7-,14-,15-,21-;/m0./s1
SMILES Cl.CN(C)[C@H]1[C@@H]2C[C@@H]3[C@H](O)c4c(Cl)ccc(O)c4C(=O)C3=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N
IUPAC Name (4S,4aS,5aS,6S,12aS)-7-chloro-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-1,11-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrochloride
Molecular Weight (g/mol) 501.31
Quantity 250 mg
Formula Weight 500.0753 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.