missing translation for 'onlineSavingsMsg'
Learn More

Demeclocycline Hydrochloride, LoGiCal

Product Code. 30460925
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460925 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460925 Supplier LoGiCal Supplier No. LGCFOR0636.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Demeclocycline Hydrochloride
CAS 64-73-3
Analyte or Component Names Demeclocycline Hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C21 H21 Cl N2 O8 . Cl H
Certifications/Compliance ISO 9001
Synonym Chlortetracycline Hydrochloride Imp. B (EP), Chlortetracycline Imp. B (EP), Demeclocycline Hydrochloride, Chlortetracycline Hydrochloride Imp. B (EP) as Hydrochloride, 7-Chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride, (4S,4aS,5aS,6S,12aS)-7-Chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride, [4S-(4alpha,4aalpha,5aalpha,6beta,12aalpha)]-7-Chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride, 6-Demethyl-7-chlorotetracycline hydrochloride, 7-Chloro-6-demethyltetracycline hydrochloride, Clortetrin, Declomycin, Declomycin hydrochloride, Demeclocycline hydrochloride, Demethylchlorotetracycline hydrochloride, Demetraciclina, Detravis, Ledermycin hydrochloride, Meciclin, Chlortetracycline Hydrochloride Impurity B as Hydrochloride, Chlortetracycline Impurity B as Hydrochloride
InChI Formula InChI=1S/C21H21ClN2O8.ClH/c1-24(2)14-7-5-6-10(16(27)12-9(25)4-3-8(22)11(12)15(6)26)18(29)21(7,32)19(30)13(17(14)28)20(23)31;/h3-4,6-7,14-15,25-26,28-29,32H,5H2,1-2H3,(H2,23,31);1H/t6-,7-,14-,15-,21-;/m0./s1
Solution Type Neat
SMILES Cl.CN(C)[C@H]1[C@@H]2C[C@@H]3[C@H](O)c4c(Cl)ccc(O)c4C(=O)C3=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N
IUPAC Name (4S,4aS,5aS,6S,12aS)-7-chloro-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-1,11-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 501.31
Quantity 10 mg
Formula Weight 500.0753 g/mol
Show More Show Less
Product Title
Select an issue

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.