missing translation for 'onlineSavingsMsg'
Learn More

Dembrexine Hydrochloride, TRC

Product Code. 30381462
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381462 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381462 Supplier Toronto Research Chemicals Supplier No. TRCD53200050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dembrexine Hydrochloride
CAS 52702-51-9
Molecular Formula C13 H17 Br2 N O2 . Cl H
Quantity 50 mg
Synonym 3,5-Dibromo-2-hydroxy-N-(trans-4-hydroxycyclohexyl)benzylamine Hydrochloride, N-(2-Hydroxy-3,5-dibromobenzyl)-trans-4-hydroxycyclohexylamine Hydrochloride
InChI Formula InChI=1S/C13H17Br2NO2.ClH/c14-9-5-8(13(18)12(15)6-9)7-16-10-1-3-11(17)4-2-10;/h5-6,10-11,16-18H,1-4,7H2;1H/t10-,11-;
SMILES Cl.O[C@@H]1CC[C@H](CC1)NCc2cc(Br)cc(Br)c2O
IUPAC Name 2,4-dibromo-6-[[(4-hydroxycyclohexyl)amino]methyl]phenol;hydrochloride
Molecular Weight (g/mol) 415.55
Formula Weight 412.9393
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.