missing translation for 'onlineSavingsMsg'
Learn More

Dematin Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18331316
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18331316 25 μg 25µL
18383412 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18331316 Supplier Novus Biologicals Supplier No. NBP31782925UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Dematin Polyclonal antibody specifically detects Dematin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dematin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000 - 1:10000
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias dematin, DMTFLJ78462, Erythrocyte membrane protein band 4.9, erythrocyte membrane protein band 4.9 (dematin), FLJ98848
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: DDDSGEEMKALRERQREELSKVTSNLGKMILKEEMEKSLPIRRKTRSLPDRTPFHTSLHQGT
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2039
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.