Learn More
DEM1/EXO5 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25991-1-AP-150UL
Description
The DEM1/EXO5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human DEM1/EXO5. This antibody recognizes human, mouse antigen. The DEM1/EXO5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| DEM1/EXO5 | |
| 350 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| C1orf176, DEM1, EXO5, Exonuclease V | |
| 150 μL | |
| 64789 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |