missing translation for 'onlineSavingsMsg'
Learn More

delta-Valerolactam, 98%

Product Code. 11474983
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474983 25 g 25g
11484983 100 g 100g
11494983 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474983 Supplier Thermo Scientific Alfa Aesar Supplier No. A12271.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 675-20-7 | C5H9NO | 99.13 g/mol

delta-Valerolactam is the starting material for a new route to hexahydrojulolidines, using intramolecular 1,4-cycloaddition. It is used as an intermediate in the preparation of other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
delta-Valerolactam is the starting material for a new route to hexahydrojulolidines, using intramolecular 1,4-cycloaddition. It is used as an intermediate in the preparation of other chemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 675-20-7
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00006037
InChI Key XUWHAWMETYGRKB-UHFFFAOYSA-N
Synonym 2-piperidone, 2-piperidinone, delta-valerolactam, valerolactim, 5-pentanolactam, piperidinone, piperidon, piperidone, alpha-piperidone, piperidone-2
PubChem CID 12665
ChEBI CHEBI:77761
IUPAC Name piperidin-2-one
SMILES O=C1CCCCN1

Specifications

Melting Point 35°C to 39°C
Boiling Point 255°C to 257°C
Flash Point 147°C (296°F)
Quantity 25 g
Beilstein 106434
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 99.13
Percent Purity ≥98%
Chemical Name or Material delta-Valerolactam
compliance-icons
Product Identifier
  • delta-Valerolactam
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Piperidinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.