missing translation for 'onlineSavingsMsg'
Learn More

delta-Tetradecanolactone, 98%

Product Code. 11496536
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496536 25 g 25g
11406546 100 g 100g
11416546 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496536 Supplier Thermo Scientific Alfa Aesar Supplier No. B24794.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2721-22-4 | C14H26O2 | 226.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2721-22-4
Molecular Formula C14H26O2
Molecular Weight (g/mol) 226.36
MDL Number MFCD00059809
InChI Key SKQYTJLYRIFFCO-UHFFFAOYNA-N
Synonym delta-tetradecalactone, 5-tetradecanolide, 2h-pyran-2-one, tetrahydro-6-nonyl, delta-tetradecanolide, delta-tetradecanolactone, tetrahydro-6-nonyl-2h-pyran-2-one, fema no. 3590, 6-nonyltetrahydro-2h-pyran-2-one, delta-myristolactone, 5-hydroxytetradecanoic acid delta-lactone
PubChem CID 520296
IUPAC Name 6-nonyloxan-2-one
SMILES CCCCCCCCCC1CCCC(=O)O1

Specifications

Melting Point 130°C to 135°C
Density 0.935
Flash Point >110°C (230°F)
Quantity 25 g
Formula Weight 226.36
Percent Purity 98%
Chemical Name or Material delta-Tetradecanolactone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.