missing translation for 'onlineSavingsMsg'
Learn More
Learn More
delta-Sarcoglycan Antibody (19G8), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42662JF646
This item is not returnable.
View return policy
Description
delta-Sarcoglycan Monoclonal antibody specifically detects delta-Sarcoglycan in Human samples. It is validated for Western Blot, ELISA
Specifications
| delta-Sarcoglycan | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 35 kDa dystrophin-associated glycoprotein, 35DAG, 35kD dystrophin-associated glycoprotein, CMD1LSG-delta, DAGD, delta-sarcoglycan, delta-SG, dystrophin associated glycoprotein, delta sarcoglycan, LGMD2F, MGC22567, placental delta sarcoglycan, sarcoglycan, delta (35kD dystrophin-associated glycoprotein), sarcoglycan, delta (35kDa dystrophin-associated glycoprotein), SGCDP, SGD | |
| Recombinant human delta-Sarcoglycan (57-289aa) purified from E. coli | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 6444 | |
| Store at 4°C in the dark. | |
| IgG1b κ |
| Western Blot, ELISA | |
| 19G8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction