missing translation for 'onlineSavingsMsg'
Learn More
Learn More
delta Opioid R/OPRD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£173.00 - £328.00
Specifications
| Antigen | delta Opioid R/OPRD1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18267085
|
Novus Biologicals
NB110-79877SS |
0.025 mg |
£173.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18727623
|
Novus Biologicals
NB110-79877 |
0.1 mg |
£328.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
delta Opioid R/OPRD1 Polyclonal specifically detects delta Opioid R/OPRD1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| delta Opioid R/OPRD1 | |
| Western Blot 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Membrane Trafficking and Chaperones | |
| PBS with 0.02% Sodium Azide | |
| delta opioid receptor 1, delta-type opioid receptor, D-OR-1, DOR-1, opioid receptor, delta 1, OPRD | |
| OPRD1 | |
| IgG | |
| Affinity Purified | |
| Delta Opiod Receptor |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Mouse, Rat, Human | |
| P41143 | |
| 4985 | |
| Synthetic peptide comprising residues 361-372 of the human DOR-1 protein. Reacts with mouse and rat DOR-1. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 40 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title