missing translation for 'onlineSavingsMsg'
Learn More

Delta-Octanolactone, TRC

Product Code. 30421625
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421625 2.5 g 2.5g
30418539 250 mg -
30420555 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30421625 Supplier Toronto Research Chemicals Supplier No. TRCO2397282.5G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698-76-0
Molecular Formula C8 H14 O2
Molecular Weight (g/mol) 142.2
Synonym Tetrahydro-6-propyl-2H-pyran-2-one, 5-Hydroxyoctanoic acid lactone, 5-Octanolide, δ-Propyl-δ-valerolactone
IUPAC Name 6-propyltetrahydropyran-2-one
SMILES CCCC1CCCC(=O)O1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C8H14O2/c1-2-4-7-5-3-6-8(9)10-7/h7H,2-6H2,1H3
Formula Weight 142.0994
Chemical Name or Material delta-Octalactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.