missing translation for 'onlineSavingsMsg'
Learn More

delta-Octanolactone, 97%

Product Code. 11333456
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333456 25 g 25g
11343456 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333456 Supplier Thermo Scientific Alfa Aesar Supplier No. L14303.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 698-76-0 | C8H14O2 | 142.198 g/mol

delta-Octanolactone is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
delta-Octanolactone is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698-76-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00144051
InChI Key FYTRVXSHONWYNE-UHFFFAOYSA-N
Synonym delta-octanolactone, delta-octalactone, 5-octanolide, 5-hydroxyoctanoic acid lactone, 2h-pyran-2-one, tetrahydro-6-propyl, 5-octalactone, .delta.-octalactone, tetrahydro-6-propyl-2h-pyran-2-one, delta-octanolide, fema no. 3214
PubChem CID 12777
IUPAC Name 6-propyloxan-2-one
SMILES CCCC1CCCC(=O)O1

Specifications

Melting Point -14°C
Density 1.002
Boiling Point 238°C
Flash Point 125°C (257°F)
Refractive Index 1.455
Quantity 25 g
Beilstein 111515
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 142.2
Percent Purity 97%
Chemical Name or Material delta-Octanolactone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.