missing translation for 'onlineSavingsMsg'
Learn More

delta-Dodecanolactone, 98%

Product Code. 11333516
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333516 25 g 25g
11343516 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333516 Supplier Thermo Scientific Alfa Aesar Supplier No. L14406.14
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 713-95-1 | C12H22O2 | 198.306 g/mol

delta-Dodecanolactone is used as a :flavoring agent. It is used in daily chemical essence. It is also used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
delta-Dodecanolactone is used as a :flavoring agent. It is used in daily chemical essence. It is also used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Handle and store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 713-95-1
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00006651
InChI Key QRPLZGZHJABGRS-UHFFFAOYSA-N
Synonym delta-dodecalactone, 5-dodecanolide, delta-dodecanolactone, dodecan-5-olide, 6-heptyltetrahydro-2h-pyran-2-one, 5-dodecalactone, 2h-pyran-2-one, 6-heptyltetrahydro, dodecanolide-1,5, 5-hydroxydodecanoic acid lactone, n-heptyl-delta-valerolactone
PubChem CID 12844
IUPAC Name 6-heptyloxan-2-one
SMILES CCCCCCCC1CCCC(=O)O1

Specifications

Melting Point -7°C
Density 0.95
Boiling Point 288°C
Flash Point 165°C (329°F)
Refractive Index 1.46
Quantity 25 g
Beilstein 1282749
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 198.3
Percent Purity 98%
Chemical Name or Material delta-Dodecanolactone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.