missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ delta Catenin Monoclonal Antibody (K29002_1E2), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA555807

 View more versions of this product

Product Code. 30212696

  • £416.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: YIRSALRQEK ALSAIADLLT NEHERVVKAA SGALRNLAVD ARNKELIGKH AIPNLVKNLP GGQQNSSWNF SEDTVISILN TINEVIAENL EAAKKLRETQ GIEKLVLINK SGNRSEKEVR AAALVLQTIW GYKELRKPLE KEGWKKSDFQ VNLNNASRSQ SSHSYDDSTL PLIDRNQKSD KKPDREEIQM SNMGSNTKSL DNNYSTPNER GDHNRTLDRS GDLGDMEPLK GTTPLMQDEG

Delta 1 Catenin is an efficient tyrosine kinase substrate implicated both in cell transformation by SRC and in ligand-induced receptor signaling through the EGF, PDGF, CSF-1 and ERBB2 receptors. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

delta Catenin
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
O60716
CTNND1
Recombinant human CTNND1 (immunogen range is 701-940).
50 μL
Primary
Human
Antibody
IgG2b
Immunoprecipitation, Western Blot
K29002_1E2
Unconjugated
CTNND1
AA409437; AU019353; cadherin-associated Src substrate; Cadherin-associated Src substrate (CAS); CAS; Catenin (cadherin associated protein) delta 1; catenin (cadherin associated protein), delta 1; catenin (cadherin-associated protein), delta 1; catenin delta 1; catenin delta-1; catenin src; catenin, delta 1; Catns; CTNND; Ctnnd1; hypothetical protein; Kiaa0384; mKIAA0384; p120; p120 catenin; P120 CTN; P120 p120; P120 P120CTN; p120(CAS); p120(ctn); P120CAS; p120-catenin; P120CTN
Mouse
Protein A
RUO
1500
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.