Learn More
Invitrogen™ delta Catenin Monoclonal Antibody (K29002_1E2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556187
Description
Sequence of this protein is as follows: YIRSALRQEK ALSAIADLLT NEHERVVKAA SGALRNLAVD ARNKELIGKH AIPNLVKNLP GGQQNSSWNF SEDTVISILN TINEVIAENL EAAKKLRETQ GIEKLVLINK SGNRSEKEVR AAALVLQTIW GYKELRKPLE KEGWKKSDFQ VNLNNASRSQ SSHSYDDSTL PLIDRNQKSD KKPDREEIQM SNMGSNTKSL DNNYSTPNER GDHNRTLDRS GDLGDMEPLK GTTPLMQDEG
Delta 1 Catenin is an efficient tyrosine kinase substrate implicated both in cell transformation by SRC and in ligand-induced receptor signaling through the EGF, PDGF, CSF-1 and ERBB2 receptors. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties.
Specifications
| delta Catenin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| O60716 | |
| CTNND1 | |
| Recombinant human CTNND1 (immunogen range is 701-940). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| K29002_1E2 | |
| Unconjugated | |
| CTNND1 | |
| AA409437; AU019353; cadherin-associated Src substrate; Cadherin-associated Src substrate (CAS); CAS; Catenin (cadherin associated protein) delta 1; catenin (cadherin associated protein), delta 1; catenin (cadherin-associated protein), delta 1; catenin delta 1; catenin delta-1; catenin src; catenin, delta 1; Catns; CTNND; Ctnnd1; hypothetical protein; Kiaa0384; mKIAA0384; p120; p120 catenin; P120 CTN; P120 p120; P120 P120CTN; p120(CAS); p120(ctn); P120CAS; p120-catenin; P120CTN | |
| Mouse | |
| Protein A | |
| RUO | |
| 1500 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.