missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ delta Catenin Monoclonal Antibody (15D2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ 339600
This item is not returnable.
View return policy
Description
delta Catenin Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ELISA
Delta 1 Catenin is an efficient tyrosine kinase substrate implicated both in cell transformation by SRC and in ligand-induced receptor signaling through the EGF, PDGF, CSF-1 and ERBB2 receptors. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties.
Specifications
| delta Catenin | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.1% sodium azide | |
| O60716, P30999 | |
| CTNND1 | |
| Recombinant fusion protein derived from the C-terminus of murine p120ct. | |
| RUO | |
| 12388, 1500, 311163, 483489 | |
| -20°C | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 15D2 | |
| Unconjugated | |
| CTNND1 | |
| AA409437; AU019353; cadherin-associated Src substrate; Cadherin-associated Src substrate (CAS); CAS; Catenin (cadherin associated protein) delta 1; catenin (cadherin associated protein), delta 1; catenin (cadherin-associated protein), delta 1; catenin delta 1; catenin delta-1; catenin src; catenin, delta 1; Catns; CTNND; Ctnnd1; hypothetical protein; Kiaa0384; mKIAA0384; p120; p120 catenin; P120 CTN; P120 p120; P120 P120CTN; p120(CAS); p120(ctn); P120CAS; p120-catenin; P120CTN | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction