Learn More
Polycarbin Delta Carbin Program for PPE Recycling (5-pack)
GREENER_CHOICE
Product Code. 31702908
Change view
Click to view available options
Product Type:
Recycling and Sustainability
Unit Size:
Pack of 5
Product Code. Product Type unitSize
31702908 Recycling and Sustainability Pack of 5
1 options
Product Code. 31702908 Supplier Polycarbin Supplier No. PR010401UK
Edge
Closed-loop mailback recycling for non-hazardous lab PPE supporting waste diversion and sustainability tracking.
  • Closed-Loop Recycling for Laboratory PPE: Enables laboratories to recycle a wide range of non-infectious, non-hazardous PPE, offering a sustainable alternative to traditional disposal methods while promoting circularity.
  • Simple Mailback Process: Each kit includes five (5) 61-liter (16-gallon) collection containers with prepaid return labels, allowing easy placement across facilities and direct return to Polycarbin for recycling.
  • Universal PPE Compatibility: Designed to collect common laboratory PPE such as disposable lab coats, gowns, masks, hairnets, and shoe covers for streamlined recovery.
  • Safe and Compliant: Accepts only non-hazardous, non-infectious PPE. Excludes items with metal clips, zippers, cloth, or paper and those contaminated with hazardous, pathological, or infectious waste.
  • Verified Impact Tracking: Includes access to Polycarbin's Carbin Counter™ platform, providing ISO 14040/14044-compliant, third-party verified reporting on waste diversion, carbon reduction, and material recovery.
  • Supports ESG and Scope 3 Goals: Recovered PPE is processed and reintegrated into Polycarbin’s low-carbon lab products, supporting sustainability and circular procurement initiatives.
  • Cost and Carbon Savings: Diverts PPE waste from landfill and incineration, helping labs lower disposal expenses and reduce their carbon footprint.
  • Compatible with Laboratory Standards: Integrates smoothly into regulated laboratory environments and existing waste segregation workflows, ensuring full compliance.
  • Convenient Configuration: Includes five (5) 61-liter receptacles ideal for distributed placement across lab spaces and common areas for maximum efficiency.
  • Proven Circular Solution: Ensures collected PPE materials are repurposed into durable, low-carbon lab products-closing the loop on laboratory PPE waste.
Specifications
Product Type Recycling and Sustainability