missing translation for 'onlineSavingsMsg'
Learn More

delta 2 Catenin Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18332214 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18332214 20 μL 20µL
18377844 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18332214 Supplier Novus Biologicals Supplier No. NBP30382520UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

delta 2 Catenin Polyclonal specifically detects delta 2 Catenin in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen delta 2 Catenin
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias catenin (cadherin-associated protein), delta 2 (neural plakophilin-relatedarm-repeat protein), catenin delta-2, Delta-catenin, GT24T-cell delta-catenin, Neural plakophilin-related ARM-repeat protein, Neurojungin, NPRAPneurojungin
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 966-1225 of human delta 2 Catenin (NP_001323.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 1501
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.