missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach™ Delivery Tubes for Digital Titrator
Description
Delivery tubes attach to the Titrator Cartridge and is used to precisely add titrant to the sample one drop at a time until the color changes.
Specifications
Specifications
| Format | Straight, with J Hook |
| For Use With (Application) | Titration |
| For Use With (Equipment) | Digital Titrator, Titrator Cartridge |
| Quantity | 5 Pack |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?