missing translation for 'onlineSavingsMsg'
Learn More

Delgocitinib, MedChemExpress

Product Code. 16450597
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16450597 50 mg 50mg
16446987 5 mg 5mg
16445196 100 mg 100mg
16443016 25 mg 25mg
16434268 10 mg 10mg
16421129 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16450597 Supplier Medchem Express Supplier No. HY109053
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Delgocitinib (JTE-052) is a specific JAK inhibitor with IC50s of 2.8, 2.6, 13 and 58 nM for JAK1, JAK2, JAK3 and Tyk2, respectively.

Delgocitinib (JTE-052) is a specific JAK inhibitor with IC50s of 2.8, 2.6, 13 and 58 nM for JAK1, JAK2, JAK3 and Tyk2, respectively.

Specifications

Chemical Name or Material Delgocitinib
CAS 1263774-59-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H18N6O
Quantity 50 mg
Synonym JTE-052
Solubility Information DMSO : ≥ 58 mg/mL (186.89 mM)
SMILES O=C(N(C[C@@H]1C)[C@]21CN(C3=C(C=CN4)C4=NC=N3)CC2)CC#N
Molecular Weight (g/mol) 310.35
Formula Weight 310.35
Percent Purity 99.75%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.