missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Deleted In Lymphocytic Leukemia, 7 Polyclonal specifically detects Deleted In Lymphocytic Leukemia, 7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Deleted In Lymphocytic Leukemia, 7 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | Q6UYE1 |
| Gene Alias | Deleted In Lymphocytic Leukemia 7, DLEU7, LEU7, Leukemia-Associated Protein 7 |
| Gene Symbols | DLEU7 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?