missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Delapril Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Delapril Hydrochloride |
| CAS | 83435-67-0 |
| Molecular Formula | C26 H32 N2 O5 . Cl H |
| Quantity | 25 mg |
| Synonym | Delapril Hydrochloride, N-[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]-L-alanyl-N-(2,3-dihydro-1H-inden-2-yl)glycine Monohydrochloride, (S)-N-(2,3-Dihydro-1H-inden-2-yl)-N-[N-[1-(ethoxycarbonyl)-3-phenylpropyl]-L-alanyl]glycine Monohydrochloride, Adecut, CV 3317, Cupressin, REV 6000A |
| InChI Formula | InChI=1S/C26H32N2O5.ClH/c1-3-33-26(32)23(14-13-19-9-5-4-6-10-19)27-18(2)25(31)28(17-24(29)30)22-15-20-11-7-8-12-21(20)16-22;/h4-12,18,22-23,27H,3,13-17H2,1-2H3,(H,29,30);1H/t18-,23-;/m0./s1 |
| SMILES | Cl.CCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N(CC(=O)O)C2Cc3ccccc3C2 |
| IUPAC Name | 2-[2,3-dihydro-1H-inden-2-yl-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]amino]acetic acid;hydrochloride |
| Molecular Weight (g/mol) | 489.004 |
| Formula Weight | 488.208 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?