missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Delangin Isoform B Monoclonal Antibody (KT55)
GREENER_CHOICE

Product Code. 11570402
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11570402 100 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11570402 Supplier Invitrogen™ Supplier No. MA172535

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Delangin Isoform B Monoclonal Antibody for IHC (P)

Probably plays a structural role in chromatin. Involved in sister chromatid cohesion, possibly by interacting with the cohesin complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Delangin Isoform B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone KT55
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene NIPBL
Gene Accession No. Q6KC79
Gene Alias 4921518A06Rik; 4933421G18Rik; C79399; CDLS; CDLS1; Delangin; delangin homolog; ID; IDN3; IDN3-B; NIPBL; NIPBL, cohesin loading factor; Nipped-B homolog; Nipped-B homolog (Drosophila); nipped-B-like protein; Scc2; SCC2 homolog; sister chromatid cohesion 2 homolog
Gene Symbols NIPBL
Host Species Rat
Immunogen Delangin peptide coupled to BSA.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25836
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.